Skip to Content
Merck
All Photos(2)

Key Documents

857323P

Avanti

16:0 Cyclic LPA

1-palmitoyl-sn-glycero-2,3-cyclic-phosphate (ammonium salt), powder

Synonym(s):

1-hexadecanoyl-sn-glycero-2,3-cyclic-phosphate (ammonium salt); cPA; cLPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H40NO6P
CAS Number:
Molecular Weight:
409.50
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (857323P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857323P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=P1([O-])O[C@H](COC(CCCCCCCCCCCCCCC)=O)CO1.[NH4+]

InChI key

NATIUIORRLVXDB-GMUIIQOCSA-N

General description

Cyclic phosphatidic acid (cPA) is a naturally occurring analog of the growth factor-like phospholipid mediator, lysophosphatidic acid (LPA). The sn-2 hydroxy group of CPA forms a 5-membered ring with the sn-3 phosphate. cPA affects numerous cellular functions, including anti-mitogenic regulation of the cell cycle, induction of stress fiber formation, inhibition of tumor cell invasion and metastasis, and regulation of differentiation and survival of neuronal cells.
Interestingly, many of these cellular responses caused by cPA oppose those of LPA despite the activation of apparently overlapping receptor populations.

Application

16:0 Cyclic LPA may be used for the extraction recovery of cyclic phosphatidic acid (cPA) at different pH conditions using Bligh-Dyer method.

Packaging

5 mL Amber Glass Screw Cap Vial (857323P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative determination of cyclic phosphatidic acid in human serum by LC/ESI/MS/MS
Shan L, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 862(1-2), 161-167 (2008)
Cyclic phosphatidic acid elicits neurotrophin-like actions in embryonic hippocampal neurons.
Fujiwara Y, et al.
Journal of Neurochemistry, 87, 1272-1283 (2003)
Yuko Fujiwara et al.
Journal of neurochemistry, 87(5), 1272-1283 (2003-11-19)
Cyclic phosphatidic acid (cPA; 1-acyl-sn-glycerol-2,3-cyclic phosphate) is an analog of the growth factor-like phospholipid mediator lysophosphatidic acid (LPA). As brain tissue is the richest source of cPA we tested its effects on hippocampal neurons from day 16/17 embryonic rat cultured
D J Fischer et al.
Molecular pharmacology, 54(6), 979-988 (1998-12-18)
Lysophosphatidic acid (LPA), plasmalogen-glycerophosphate (alkenyl-GP) and, cyclic-phosphatidic acid (cyclic-PA) are naturally occurring phospholipid growth factors (PLGFs). PLGFs elicit diverse biological effects via the activation of G protein-coupled receptors in a variety of cell types. In NIH3T3 fibroblasts, LPA and alkenyl-GP
K Murakami-Murofushi et al.
Cell structure and function, 18(5), 363-370 (1993-10-01)
The unique Physarum lysophosphatidic acid, PHYLPA, having a cyclopropane in the fatty acid moiety and a cyclic phosphate at C-2 and C-3 positions of the glycerol, inhibited proliferation of human fibroblast cells, TIG-3 and TIG-7, which were cultured in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service