Skip to Content
Merck
All Photos(2)

Key Documents

855575C

Avanti

14:0 Lyso PC

1-myristoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-tetradecanoyl-sn-glycero-3-phosphocholine; PC(14:0/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H46NO7P
CAS Number:
Molecular Weight:
467.58
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (855575C-25mg)
pkg of 1 × 20 mL (855575C-500mg)
pkg of 2 × 20 mL (855575C-1g)
pkg of 2 × 4 mL (855575C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855575C

concentration

10 mg/mL (855575C-25mg)
25 mg/mL (855575C-1g)
25 mg/mL (855575C-200mg)
25 mg/mL (855575C-500mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](O)COC(=O)CCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C22H46NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-22(25)28-19-21(24)20-30-31(26,27)29-18-17-23(2,3)4/h21,24H,5-20H2,1-4H3/t21-/m1/s1

InChI key

VXUOFDJKYGDUJI-OAQYLSRUSA-N

Application

14:0 Lyso PC has been used as a lipid standard in the optimization of the liquid chromatography-mass spectrometry (LC-MS) method conditions. It has also been used to engineer collagen hydrogels and examine the in vitro release profiles of fluorescently labeled or computed tomography (CT) contrast biomolecules.

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (855575C-1g)
30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (855575C-500mg)
5 mL Clear Glass Sealed Ampule (855575C-200mg)
5 mL Clear Glass Sealed Ampule (855575C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Norimichi Nagano et al.
Biological & pharmaceutical bulletin, 42(11), 1926-1935 (2019-11-07)
Elevated intraocular pressure (IOP) is the major cause of glaucoma, which is the second leading cause of blindness. However, current glaucoma treatments cannot completely regulate IOP and progression of glaucoma. Our group recently found that autotaxin (ATX) activity in human
Baasanjav Uranbileg et al.
Scientific reports, 9(1), 16578-16578 (2019-11-14)
Cauda equina compression (CEC) is a major cause of neurogenic claudication and progresses to neuropathic pain (NP). A lipid mediator, lysophosphatidic acid (LPA), is known to induce NP via the LPA1 receptor. To know a possible mechanism of LPA production
Julia Koehler et al.
Biochemistry, 49(33), 7089-7099 (2010-07-30)
There has been a renewal of interest in interactions of membrane proteins with detergents and lipids, sparked both by recent results that illuminate the structural details of these interactions and also by the realization that some experimental membrane protein structures
Christoph Göbl et al.
The journal of physical chemistry. B, 114(13), 4717-4724 (2010-03-17)
The interaction with biological membranes is of functional importance for many peptides and proteins. Structural studies on such membrane-bound biomacromolecules are often carried out in solutions containing small membrane-mimetic assemblies of detergent molecules. To investigate the influence of the hydrophobic
Reinhard Zeisig et al.
Cancer gene therapy, 10(4), 302-311 (2003-04-08)
To improve liposomal gene transfer we investigated the influence of membrane-interacting alkylphospholipids (APLs) on gene transfer efficiency in vitro and in vivo using the LacZ reporter gene and the cytosine deaminase (CD) suicide gene. Liposomes were first optimized concerning the

Articles

The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service