Skip to Content
Merck
All Photos(2)

Key Documents

850376P

Avanti

18:1 (Δ9-Trans) PC

1,2-dielaidoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

1,2-di-(9E-octadecenoyl)-sn-glycero-3-phosphocholine; PC(18:1(9E)/18:1(9E))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H84NO8P
CAS Number:
Molecular Weight:
786.11
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 2 × 100 mg (850376P-200mg)
pkg of 1 × 25 mg (850376P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850376P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C/CCCCCCCC)=O)COC(CCCCCCC/C=C/CCCCCCCC)=O)=O

InChI

1S/C44H84NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h20-23,42H,6-19,24-41H2,1-5H3/b22-20+,23-21+

InChI key

SNKAWJBJQDLSFF-DQPVQCHKSA-N

General description

18:1 (Δ9-Trans) PC (1,2-dielaidoyl-sn-glycero-3-phosphocholine) is a lipid derivative having a phosphocholine backbone. It also has two 18 carbon fatty acid chains, each having one trans double bond at the 9th carbon. It is an isomeric glycerophosphocholine.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

18:1 (Δ9-Trans) PC (1,2-dielaidoyl-sn-glycero-3-phosphocholine) may be used:
  • in the preparation of liposomes/vesicles to examine peptide-mediated leakage
  • as a standard phospholipid and a model compound to study the effect of several matrix-assisted laser desorption/ionization (MALDI) matrixes on the degree of the fragmentation of different phospholipids
  • as a phosphatidylcholine standard to demonstrate the analytical utility of performing ozonolysis (LC-Oz-IM-MS)

Biochem/physiol Actions

18:1 (Δ9-Trans) PC (1,2-dielaidoyl-sn-glycero-3-phosphocholine) can help paclitaxel in forming thermodynamically stable systems. It has efficient drug penetration ability into the lipid monolayers when compared to other lipids.

Packaging

5 mL Clear Glass Sealed Ampule (850376P-200mg)
5 mL Clear Glass Sealed Ampule (850376P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junling Ren et al.
Scientific reports, 6, 34201-34201 (2016-10-01)
In this study, we have employed graphene oxide as a matrix to simultaneously and directly quantify serum nonesterified and esterified fatty acids (FAs) using matrix-assisted laser/desorption ionization-Fourier transform ion cyclotron resonance mass spectrometry (MALDI-FTICR MS). Twelve serum nonesterified FAs combined
Lingyun Zhao et al.
Journal of colloid and interface science, 285(1), 326-335 (2005-03-31)
Molecular interactions between paclitaxel, an anticancer drug, and phospholipids of various chain unsaturations and headgroup types were investigated in the present study by Langmuir film balance and differential scanning calorimetry. Both the lipid monolayer at the air-water interface and the
Rachel A Harris et al.
Analytical chemistry, 90(3), 1915-1924 (2018-01-18)
The increasing focus on lipid metabolism has revealed a need for analytical techniques capable of structurally characterizing lipids with a high degree of specificity. Lipids can exist as any one of a large number of double bond positional isomers, which
F Nicol et al.
Biophysical journal, 78(2), 818-829 (2000-02-02)
To better understand the influence of phospholipid acyl-chain composition on the formation of pores by cytotoxic amphipathic helices in biological membranes, the leakage of aqueous contents induced by the synthetic peptide GALA (WEAALAEALAE ALAEHLAEALAEALEALAA) from large unilamellar phospholipid vesicles of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service