Skip to Content
Merck
All Photos(2)

Key Documents

850373P

Avanti

24:0 PC

1,2-dilignoceroyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

1,2-ditetracosanoyl-sn-glycero-3-phosphocholine; PC(24:0/24:0) ; 110929

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H112NO8P
CAS Number:
Molecular Weight:
958.46
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 200 mg (850373P-200mg)
pkg of 1 × 25 mg (850373P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850373P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)(C)C)=O)(OC(CCCCCCCCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCCCCCCCC)=O

InChI

1S/C56H112NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-38-40-42-44-46-48-55(58)62-52-54(53-64-66(60,61)63-51-50-57(3,4)5)65-56(59)49-47-45-43-41-39-37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h54H,6-53H2,1-5H3/t54-/m1/s1

InChI key

XOAMGMFHUNHBEM-AXAMJWTMSA-N

General description

24:0 PC (1,2-dilignoceroyl-sn-glycero-3-phosphocholine) is a long-chain diacyl phospholipid. It consists of C24 saturated hydrocarbon chains.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

24:0 PC (1,2-dilignoceroyl-sn-glycero-3-phosphocholine) may be used:
  • in both short-chain micelles and longer-chain oriented multilayers and in Langmuir monolayers for structural studies of the human immunodeficiency virus 1 (HIV-1) accessory protein
  • to compare cohesion/to study the effects of lipid cohesion on the thermally induced revaporization and dissolution of microbubble condensed droplets (MCDs)
  • to study its effects on non-amyloidogenic processing of amyloid precursor protein (APP) and to study the effects of the its carbon chain length on α-secretase activity

Biochem/physiol Actions

Phosphatidylcholines (PCs) serve as a major source for eicosanoids.

Packaging

5 mL Amber Glass Screw Cap Vial (850373P-200mg)
5 mL Amber Glass Screw Cap Vial (850373P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul A Mountford et al.
Langmuir : the ACS journal of surfaces and colloids, 31(16), 4627-4634 (2015-04-09)
This study explored the thermal conditions necessary for the vaporization of superheated perfluorocarbon nanodrops. Droplets C3F8 and C4F10 coated with a homologous series of saturated diacylphosphatidylcholines were formed by condensation of 4 μm diameter microbubbles. These drops were stable at
S Zheng et al.
Biophysical journal, 80(4), 1837-1850 (2001-03-22)
Vpu is an 81 amino acid integral membrane protein encoded by the HIV-1 genome with a N-terminal hydrophobic domain and a C-terminal hydrophilic domain. It enhances the release of virus from the infected cell and triggers degradation of the virus
Marcus O W Grimm et al.
International journal of molecular sciences, 14(3), 5879-5898 (2013-03-15)
Alzheimer's disease (AD) is characterized by extracellular accumulation of amyloid-β peptide (Aβ), generated by proteolytic processing of the amyloid precursor protein (APP) by β- and γ-secretase. Aβ generation is inhibited when the initial ectodomain shedding is caused by α-secretase, cleaving
Keishi Kanno et al.
Biochimica et biophysica acta, 1771(6), 654-662 (2007-05-15)
Phosphatidylcholine transfer protein (PC-TP) is a highly specific soluble lipid binding protein that transfers phosphatidylcholine between membranes in vitro. PC-TP is a member of the steroidogenic acute regulatory protein-related transfer (START) domain superfamily. Although its biochemical properties and structure are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service