Skip to Content
Merck
All Photos(2)

Key Documents

840861P

Avanti

18:0-18:1 PA

1-stearoyl-2-oleoyl-sn-glycero-3-phosphate (sodium salt), powder

Synonym(s):

1-octadecanoyl-2--(9Z-octadecenoyl)-sn-glycero-3-phosphate (sodium salt); SOPA; PA(18:0/18:1(9Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H74O8PNa
CAS Number:
Molecular Weight:
724.96
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (840861P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840861P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C39H75O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(40)45-35-37(36-46-48(42,43)44)47-39(41)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h18,20,37H,3-17,19,21-36H2,1-2H3,(H2,42,43,44);/q;+1/p-1/b20-18-;/t37-;/m1./s1

InChI key

QTWIAVIWKTYOHY-HTNAYUJGSA-M

General description

Phosphatidic acid (PA), a phospholipid is a minor component of the phospholipid pool. It is an important constituent of cell membranes.

Application

18:0-18:1 PA (1-stearoyl-2-oleoyl-sn-glycero-3-phosphate (sodium salt)) may be used as a phosphatidic acid (PA) standard to identify the corresponding radiolabeled PA on the thin-layer chromatography (TLC). It may be also used in vesicles for experimental validation in order to identify novel phospholipids capable of inhibiting cellular membrane transportation by (ATP)-binding cassette P-glycoprotein (P-gp).

Biochem/physiol Actions

Phosphatidic acid (PA) serves as a lipid messenger and a signaling lipid.

Packaging

5 mL Clear Glass Sealed Ampule (840861P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guillermo Escalante et al.
Journal of the International Society of Sports Nutrition, 13, 24-24 (2016-06-09)
Phosphatidic acid (PA) is a lipid messenger that has been shown to increase muscle protein synthesis via signaling stimulation of the mammalian target of rapamycin (mTOR). MaxxTOR® (MT) is a supplement that contains PA as the main active ingredient but
Xavier Lucas et al.
PloS one, 8(4), e60679-e60679 (2013-04-18)
P-glycoprotein is capable of effluxing a broad range of cytosolic and membrane penetrating xenobiotic substrates, thus leading to multi-drug resistance and posing a threat for the therapeutic treatment of several diseases, including cancer and central nervous disorders. Herein, a virtual
Y Liu et al.
American journal of physiology. Lung cellular and molecular physiology, 295(3), L471-L478 (2008-07-16)
We have previously reported the participation of mitogen-activated protein, Rho, and phosphoinositide-3 (PI3) kinases in separate pathways in serotonin (5-HT)-induced proliferation of pulmonary artery smooth muscle cells (SMCs). In this study, we investigated the possible participation of phospholipase D (PLD)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service