Skip to Content
Merck
All Photos(2)

Key Documents

810285P

Avanti

TopFluor Lyso PE

Avanti Research - A Croda Brand 810285P, powder

Synonym(s):

1-(dipyrrometheneboron difluoride)undecanoyl-2-hydroxy-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H47BF2N3O7P
CAS Number:
Molecular Weight:
629.48
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (LPE; may contain up to 10% of the 2-LPE isomer)

form

powder

packaging

pkg of 1 × 1 mg (810285P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810285P

shipped in

dry ice

storage temp.

−20°C

General description

The phosphatidylethanolamine (PE) is a zwitterionic phospholipid. Its headgroup is provided with a phosphate, that is negatively charged and a free amine, that is positively charged. E is one of the widely distributed glycerophospholipid in eukaryotic cells.

Application

TopFluor Lyso PE may be used in fluorescent substrate binding and competitive binding assays.

Biochem/physiol Actions

Phosphatidylethanolamine (PE) helps to activate factor (F)X and PT, thereby stimulating thrombosis. It serves as an anticoagulant. PE is essential for the surface expression of γ-aminobutyric acid type A receptors (GABAARs) in cultured neurons.

Packaging

5 mL Amber Glass Screw Cap Vial (810285P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proteostasis maintenance of Cys-loop receptors
Advances in Protein Chemistry and Structural Biology, 103(2), 1-23 (2016)
Antibodies and Diagnostic Tests in Antiphospholipid Syndrome
Systemic Lupus Erythematosus, 268(2), 495-501 (2016)
Phosphatidylethanolamine metabolism in health and disease
International Review of Cell and Molecular Biology, 321(2), 29-88 (2016)
Lipid-protein interactions in membranes
Marsh D
Febs Letters, 268(2), 371-375 (1990)
Substrate selectivity of lysophospholipid transporter LplT involved in membrane phospholipid remodeling in Escherichia coli
Lin Y, et al.
The Journal of Biological Chemistry, 291(5), 2136-2149 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service