Skip to Content
Merck
All Photos(2)

Key Documents

810186P

Avanti

TopFluor PI(3,5)P2

Avanti Research - A Croda Brand 810186P, powder

Synonym(s):

1-oleoyl-2-{6-[4-(dipyrrometheneboron difluoride)butanoyl]amino}hexanoyl-sn-glycero-3-phosphoinositol-3,5-bisphosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H92BF2N6O20P3
CAS Number:
Molecular Weight:
1239.02
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 50 μg (with stopper and crimp cap (810186P-50ug))

manufacturer/tradename

Avanti Research - A Croda Brand 810186P

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

PI(3,5)P2 (1-oleoyl-2-{6-[4-(dipyrrometheneboron difluoride)butanoyl]amino}hexanoyl-sn-glycero-3-phosphoinositol-3,5-bisphosphate) is an endosomal lipid. It is produced by Fab1 phosphatidylinositol-3-phosphate 5-kinase from phosphatidylinositol 3-phosphate. This synthesis occurs in the lysosomal membranes.

Biochem/physiol Actions

Phosphatidylinositol phosphate lipids are associated with serval cell signalling pathways. It participates in the transport of peripheral membrane proteins along the plasma membrane. PI(3,5)P2 is essential for endosome maturation.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (810186P-50ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interactions and Translational Dynamics of Phosphatidylinositol Bisphosphate (PIP2) Lipids in Asymmetric Lipid Bilayers.
Shi X, et al.
Testing, 32(41), 1732-1741 (2016)
The phosphatidylinositol-3-phosphate 5-kinase inhibitor apilimod blocks filoviral entry and infection
Nelson EA, et al.
PLoS Neglected Tropical Diseases, 11(4), e0005540-e0005540 (2017)
The phosphatidylinositol 3, 5-bisphosphate (PI (3, 5) P2)-dependent Tup1 conversion (PIPTC) regulates metabolic reprogramming from glycolysis to gluconeogenesis
Han BK and Emr SD
The Journal of Biological Chemistry, 288(28), 20633-20645 (2013)
Xiaojun Shi et al.
Langmuir : the ACS journal of surfaces and colloids, 32(7), 1732-1741 (2016-02-02)
Phosphatidylinositol phosphate (PIP) lipids are critical to many cell signaling pathways, in part by acting as molecular beacons that recruit peripheral membrane proteins to specific locations within the plasma membrane. Understanding the biophysics of PIP-protein interactions is critical to developing
Bong-Kwan Han et al.
The Journal of biological chemistry, 288(28), 20633-20645 (2013-06-05)
Glucose/carbon metabolism is a fundamental cellular process in living cells. In response to varying environments, eukaryotic cells reprogram their glucose/carbon metabolism between aerobic or anaerobic glycolysis, oxidative phosphorylation, and/or gluconeogenesis. The distinct type of glucose/carbon metabolism that a cell carries

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service