Skip to Content
Merck
All Photos(2)

Key Documents

810118P

Avanti

Egg NBD PE

Avanti Research - A Croda Brand 810118P, powder

Synonym(s):

L-α-Phosphatidylethanolamine-N-(7-nitro-2-1,3-benzoxadiazol-4-yl) (Ammonium Salt) (Egg-Transphosphatidylated, Chicken)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H80N5O11P
CAS Number:
Molecular Weight:
898.12
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810118P-1mg)
pkg of 5 × 1 mg (810118P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810118P

shipped in

dry ice

storage temp.

−20°C

Application

Egg NBD PE may be used along with red blood cell membranes for the preparation of nano absorbents. It may also be used as a fluorescent probe to label 1, 2-dipalmitoyl-sn-glycero3-phosphocholine (DPPC) double bilayer for fluorescence microscopy studies.

Biochem/physiol Actions

L-α-Phosphatidylethanolamine-N-(7-nitro-2-1,3-benzoxadiazol-4-yl) (PE-NBD) is a fluorescent probe and is useful as a probe in lateral diffusion studies in artificial membranes.

Packaging

5 mL Amber Glass Screw Cap Vial (810118P-1mg)
5 mL Amber Glass Screw Cap Vial (810118P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Engineered biomimetic nanoabsorbent for cellular detoxification of chemotherapeutics
Nguyen T, et al.
Royal Society of Chemistry Advances, 6(39), 33003-33008 (2016)
Membranes (1981)
From supported membranes to tethered vesicles: Lipid bilayers destabilisation at the main transition
Lecuyer S and Charitat T
EPL, 75(4), 652-652 (2006)
Sagar Rayamajhi et al.
Colloids and surfaces. B, Biointerfaces, 188, 110804-110804 (2020-01-24)
Endosomal degradation of the nanoparticle is one of the major biological barriers associated with the drug delivery system. Nanoparticles are internalized in the cell via different endocytosis pathways, where they are first delivered to early endosomes which mature to the
Daniel A Peñalva et al.
Langmuir : the ACS journal of surfaces and colloids, 34(14), 4398-4407 (2018-03-16)
In rat sperm heads, sphingomyelin (SM) species that contain very long-chain polyunsaturated fatty acid (V-SM) become ceramides (V-Cer) after inducing in vitro the acrosomal reaction. The reason for such a specific location of this conversion, catalyzed by a sphingomyelinase (SMase)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service