Skip to Content
Merck
All Photos(1)

Key Documents

330678

Avanti

SM ELISA (Clear)

Avanti Research - A Croda Brand

Synonym(s):

SM ELISA, Brain Sphingomyelin ELISA Snoopers, Clear, for use in absorbance applications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

form

liquid

manufacturer/tradename

Avanti Research - A Croda Brand

packaging

1 ea of 1 × (ELISA strip packaged in foil pouch)

shipped in

wet ice

storage temp.

2-8°C

General description

Avanti Lipid Snoopers are lipid microarrays useful in semi-quantitative measurements. These snoopers are relatively stable and provide a higher sensitivity compared to the conventional enzyme-linked immunosorbent assay (ELISA) methods. Sphingomyelin (SM) is a sphingolipid present in central nervous system as a component of the myelin sheath and neuronal cells. It is synthesized from ceramide in Golgi. Acid sphingomyelinase (aSMase) degrades SM.
ELISA Snoopers are 8-well strips pre-coated with a single high-quality lipid species. They can be used to investigate protein-lipid interactions, and characterize and titrate lipid binding proteins. This format provides the following advantages:
  • quantitative lipid-protein binding studies
  • assemble a 96-well plate using only the lipids of interest for binding studies
  • flexibility - use 8 to 96 wells, and 1 to 12 lipids as needed.


The 8-well strips are available in optically clear, white, or black surfaces for use in colormetric, luminescent and fluorescent assays, respectively. Avanti Lipid Snoopers ELISA Strips allow multiple sample conditions to be probed simultaneously in a single plate making them a perfect complement to Avanti Lipid Snoopers Nitrocellulose Strips.

Biochem/physiol Actions

Sphingomyelin (SM) participates in the inflammatory process and signalling pathways. It is a crucial regulator for myelin integrity, axon maturation, cognition and brain development. The SM to ceramide ratio is crucial, as high ceremide levels are implicated in depression. Altered levels of sphingomyelin due to sphingomyelinase deficiency results in Niemann–Pick disease.

Packaging

ELISA Strip Packaged in Foil Pouch (330678-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Snoopers is a trademark of Avanti Polar Lipids, LLC

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sphingolipids: membrane microdomains in brain development, function and neurological diseases
Olsen ASB and Faergeman NJ
Open Biology, 7(5), 170069-170069 (2017)
Sphingolipid metabolism in neural cells
van ED, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1758(12), 1978-1994 (2006)
Sphingomyelin in brain and cognitive development: preliminary data
Schneider N, et al.
eNeuro (2019)
Avanti lipid tools: connecting lipids, technology, and cell biology
Sims KH, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1841(8), 1038-1048 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service