Z768316
Labeling tape
green, W × L 3/4 in. × 500 in.
Synonym(s):
green tape, label tape
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
Recommended Products
material
green
packaging
pkg of 1 roll
manufacturer/tradename
Diversified Biotech LBTP-1200
W × L
3/4 in. × 500 in.
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Strong, durable, oil & abrasion resistant, waterproof, and non-staining
- Great for general identification and color coding
- Temperature Range: -23°C to +121°C
- Accepts marking using pen, pencil, or permanent marker
- Stick to most surfaces including glass, plastic, and metal
- Repositionable, clean removable adhesive system which leaves no residue
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service