Z742947
Julabo® CORIO™ C heating immersion circulator
temperature 20-100 °C, AC/DC input 230 V AC, UK plug
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9011000
temperature measuring range
20-100 °C
W × L × H
13.2 cm × 16 cm × 36.2 cm
Looking for similar products? Visit Product Comparison Guide
General description
The CORIO C Immersion Circulator is the basic model of the CORIO circulator portfolio. The bath attachment clamp is included in delivery and facilitates mounting of the circulator on any bath tank up to 30 liters.
Your advantages
LED Temperature Display
2kW heating capacity
6 L/min Circulation capacity flow rate
Cooling coiloptional
Usable immersion depth16.6 cm
Your advantages
- Precise temperature control
- Iintegrated timer function. When the set time has elapsed, a signal sounds and the device switches off. Setting range: 0 ... 999 minutes
- For internal standard applications
- For bath tanks with a max. filling volume of 30 liters
- Bright, white, easy to read display
- Very quiet
- Easy operation
- Class I (NFL) according to DIN 12876-1
LED Temperature Display
2kW heating capacity
6 L/min Circulation capacity flow rate
Cooling coiloptional
Usable immersion depth16.6 cm
Legal Information
CORIO is a trademark of Julabo GmbH
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service