Z742918
Terg-a-zyme® enzyme detergent
package of 1.8 kg
Synonym(s):
Terg-a-zyme, Tergazyme
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
47131800
NACRES:
NB.34
Recommended Products
packaging
package of 1.8 kg
pkg of 9 × 1.8 kg
manufacturer/tradename
Alconox, Inc. 1304 (9 x 1.8 KG)
Alconox, Inc. 1304-1 (1.8 KG)
General description
Terg-a-zyme® enzyme detergent is an anionicdetergent suitable for manual and ultrasonic cleaning purposes. It facilitatesthe removal of proteinaceous soils, body fluids, tissues, and blood withoutinterfering with residues from metals, glassware, plastic, porcelain, rubber,fiberglass, and ceramic surfaces.
Application
Terg-a-zyme® enzyme detergent is suitable for:
- reverse osmosis and ultra-filtration systems.
- cleaning validation tests for lab accreditation, plant inspection approvals, and food contact surface cleaning.
- cleaning dairy equipment, pharmaceutical and cosmetics manufacturing equipment, hospital instruments, optical parts, sampling apparatus, tubing, pipes, process equipment, industrial parts, and desalinating plants, tanks, and reactors.
- cleaning soft metal surfaces made of aluminum, zinc, copper, and magnesium.
- sterilizing the body temperature detector.
- removing proteinaceous residues from implantable telemetric transponder.
- cleaning of microelectrode arrays (MEAs).
Features and Benefits
- Readily disposable as it is biodegradable
- Cost-efficient as it is concentrated
- Alternative for corrosive acids and hazardous solvents
- Free rinsing for reliable results
- Protease enzyme effectively eliminates proteinaceous soils, tissue, blood, and bodily fluids
- Suitable for FDA cleaning validations and cleaning food contact surfaces
Legal Information
Alcojet is a registered trademark of Alconox, Inc.
Detojet is a registered trademark of Alconox, Inc.
Tergazyme is a registered trademark of Alconox, Inc.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Leonie Zieglowski et al.
European surgical research. Europaische chirurgische Forschung. Recherches chirurgicales europeennes, 62(4), 271-275 (2021-06-04)
Implantable telemetric transponders for contactless measurement of physiological parameters are often used in animal-based research. After explantation, single-use devices cannot be re-implanted because of non-validated functionality and necessary re-sterilisation. This is disadvantageous because the battery life would enable a second
Cold-sensitive ventromedial hypothalamic neurons control homeostatic thermogenesis and social interaction-associated hyperthermia
Chenzang F, et al.
Cell Metabolism (2022)
David Wegrzyn et al.
Frontiers in synaptic neuroscience, 13, 637549-637549 (2021-03-13)
Perineuronal nets (PNNs) are specialized, reticular structures of the extracellular matrix (ECM) that can be found covering the soma and proximal dendrites of a neuronal subpopulation. Recent studies have shown that PNNs can highly influence synaptic plasticity and are disrupted
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service