Z742490
Generator probe for D1000 handheld homogenizer
14 mm x 130 mm saw tooth, for 50 mL tubes up to 250 mL vessels
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.61
Recommended Products
Sterilization Compatibility
autoclavable compatible
feature
14 mm x 130 mm saw tooth, for 50 mL tubes up to 250 mL vessels
packaging
pkg of 1
manufacturer/tradename
Benchmark Scientific D1000-M14
Related Categories
General description
Generator probes for Benchmark′s handheld homogenizer, the D1000.
All the generator probes are made of stainless steel and can be sterilized by autoclaving.
All the generator probes are made of stainless steel and can be sterilized by autoclaving.
Choose from one of the most recent versions:
Certificates of Analysis (COA)