Key Documents
Z742418
Q-Tube Pressure Monitor
35 SS, pack of 1 ea
About This Item
UNSPSC Code:
41121500
NACRES:
NB.51
material
316 stainless steel adapter
PTFE sleeve
packaging
pack of 1 ea
manufacturer/tradename
Q Labtech 100-PRMSET-35SS
parameter
180 psi max. pressure
General description
The parts with direct chemical contact are made of 316 stainless steel, PTFE, Simriz, or glass.
Pressure Monitor Set-35-SS/100-PRMSET-N35SS contains all items needed for immediate use (316 stainless steel Adapter and matched PTFE sleeve, 1 pressure gauge, 1 pressure tube-35 mL, 10 Septa-35 mL-180 psi, etc.).
Legal Information
Q-Tube is a trademark of Q Labtech LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)