Key Documents
Z682462
Ace vapor duct tube, for condenser "A" assembly
joint: ST/NS 24/40, L 160 mm, fits Büchi® models R-114/R-144
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Joint
joint: ST/NS24/40
manufacturer/tradename
Büchi equivalent, 32336
L
160 mm
Related Categories
General description
The vapor duct tube accessories are for Büchi® condenser "A" assemblies, and will fit either Büchi evaporator models R-210/R-215 or R-114/R-144.
Combi-clips: not included.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)