Key Documents
Z682276
Ace rotary evaporator condenser complete "CR" assembly
glass, fits Büchi® models R-200/R-205 and R-114/R-144
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
glass
manufacturer/tradename
Büchi equivalent, 11511
Related Categories
General description
This "CR" condenser assembly is similar to the "C" assembly except that the bottom joint is 45/40 inner to fit into a separate distribution head. The complete "CR" assembly consists of the main glass outer trap body, glass cold trap inner body, PTFE sealing ring with Viton® O-rings, PTFE lid and hose barbs with GL threaded caps.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)