Skip to Content
Merck
All Photos(1)

Key Documents

Z678368

Finnpipette® F2 variable volume pipette

Synonym(s):

Finnpipette F2, Variable Volume Pipette

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

feature

color-coded green
electronic: no
single-channel

packaging

pack of 1 ea

manufacturer/tradename

Finn 4642100

volume

0.5-5 mL

Looking for similar products? Visit Product Comparison Guide

General description

Finnpipette F2 has a light pipetting action, is easy to use and is fully autoclavable. Accuracy and precision are the most vital aspect of any pipette so the Finnpipette F2 was developed with Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.

  • Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
  • Big display with white numbers on black background for increased visibility
  • Double action pipetting button with very light and smooth pipetting action
  • Fully autoclavable
  • Super blow-out piston with volumes of 50μl and below

Compatibility

Uses Finntip 5mL

Legal Information

Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service