Key Documents
Z677574
Lapren latex/chloroprene flock lined gloves
size XL
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
material
green (chloroprene, natural latex)
size
XL
feature
CE compliant
disposable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
packaging
pack of 100 ea
manufacturer/tradename
KCL 706 - 10
KCL 706
L × thickness
310 mm × 0.6 mm
General description
Lapren incorporates a high quantity of polychloroprene into latex to improve chemical resistance. A mixture of raw materials ensures a high elasticity as well as good tensile strength and tear resistance. The palm grip surface ensures a firm grip even in wet working environments without loss of the high degree of sensitivity.
With cotton flock lining, stretched cuff and anatomical design, this product meets all requirements of comfort and hygiene.
  • Ergonomic fit and high elasticity ensures good sensitivity
  • Profiled palm for firm grip when wet
  • High mechanical resistance
  • Cotton flock lining improves comfort
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea
  • CE compliant
Legal Information
Lapren is a trademark of KCL GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)