Z654345
Pouring rings for Duran® laboratory bottles
GL 45 neck thread, colorless
Synonym(s):
pouring ring
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
colorless
manufacturer/tradename
DURAN®
Schott
parameter
-196-260 °C temp. stability
fitting
GL 45 neck thread
Related Categories
General description
Thanks to its highly resistant materials, the premium cap is optimally suited to utilize in processes which are both chemically and thermally demanding. Due to its high temperature resistance, it is ideal for demanding processes, such as hot-air / dry-heat sterilisation and depyrogenation at 200 °C. Due to the PTFE-coated silicone cap liner, the premium cap provides a reliable and tight seal. Therefore it is ideal for the long term storage and transport of sensitive products. The matching PFA pouring ring permits drip-free pouring is available separately. Replacement PTFE faced silicone cap liners (platinum-cured silicone) are available
Features and Benefits
Optional pouring rings prevent drips when pouring from Duran bottles.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)