Z648175
Tissue capture pen
one pen for two applications, pkg of 1 ea
Synonym(s):
histopathology pen
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
MA.01
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
22310
Related Categories
General description
INSTRUCTIONS FOR USE:
- Remove the cap. Turn the pen over (tip down) and put tip on the glass slide
- Coat an appropriate area of the slide with Tissue Capture liquid
- Pickup the section on the coated area of the slide
- Deparaffinize tissue sections and hydrate to water
- Wipe away excess liquid around the sections on the glass slide
- If using the PAP pen encircle the tissue sections or draw lines on both sides of the section with Liquid Blocker and let dry 10-15 seconds
- You are now ready to begin your labeling protocol
Features and Benefits
- Provides a sticky membrane on slide
- Prevents the tissue from moving, wrinkling or falling off the slide
- Flattens the section and eliminates the need for hot water extension
- Suitable for microwave protocols
- Use with Liquid Blocker Pen for small scale cultures
- Suitable for in-situ hybridization
- Stable up to 120°
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service