Z646199
Spinbar® restrainer
retrieve magnetic stirring bars from vessels, easily, pkg of 1 ea
All Photos(1)
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F37786-1000
Related Categories
General description
Easily retrieve magnetic stirring bars from vessels with the handheld Spinbar Restrainer. This restrainer stops losses of magnetic stirring bars headed down lab drains and eliminates the splashing hazard when pouring out of mixing containers. The tool fits comfortably in one hand and when placed next to the outside of a mixing container, readily attracts and holds stirring bars in place.
As containers are emptied, the stirring bar is restrained and can be safely removed from the vessel. The Spinbar Restrainer body is molded of easy to clean high impact polystyrene. It contains a powerful magnet that attracts and holds magnetic stirring bars up to 80 mm long.
As containers are emptied, the stirring bar is restrained and can be safely removed from the vessel. The Spinbar Restrainer body is molded of easy to clean high impact polystyrene. It contains a powerful magnet that attracts and holds magnetic stirring bars up to 80 mm long.
Legal Information
Spinbar is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)