Key Documents
Z568724
QianCap safety cap system for Ace pressure tubes and bottles
QianCap-15, fits no. 15 internal thread
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
manufacturer/tradename
Q Labtech
volume
4 oz , catch bottle
General description
QianCap is a safety cap with an automatic pressure release and reseal mechanism which can prevent pressure bottles and tubes with Ace-Threds #15 and #25 from explosion due to excessive internal pressure. These systems contain all items needed for immediate use on pressure tubes and bottles with internal threads. QianCap is available in these three configurations:
1. QianCap safety cap
2. QianCap safety cap with 0-300 psig pressure gauge
3. QianCap safety cap gas purging set, with 0-300 psig pressure gauge
Components in all configurations
  • One PTFE Q-adapter
  • One Simriz® O-ring
  • Ten septa - 12 mL
  • One pressure adapter-60
  • One cap
  • One needle adapter
  • One deflected point needle
  • One catch bottle
Legal Information
QianCap is a trademark of Q Labtech LLC
Simriz is a registered trademark of Carl Freudenburg KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)