Z548065
Aldrich® separatory funnel with PTFE stopcock
capacity 50 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
conical
capacity
50 mL
stopcock bore
2 mm
stopper size
14/20
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service