Z512141
Serrated natural rubber septa
white rubber, bottom O.D. 23 mm, fits neck I.D., 20.5 mm, ST/NS: 24/40
Synonym(s):
Natural Rubber Seal
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
white rubber
usage
ST/NS: 24/40
feature
fits neck I.D. 20.5 mm
packaging
package of 100
bottom O.D.
23 mm
Looking for similar products? Visit Product Comparison Guide
General description
Natural rubber septa/sleeve stoppers are used as temporary closures in chemistry, pharmaceuticals, research and development, and biology. These commercially available septa are developed for general applications and are mainly used in labs as closures for glassware, tubes, and cannulation of air-sensitive chemicals.
Closure has a turnover flange molded to grip outside of container neck, promoting a double seal in conjunction with annular serrations on a hollow plug. Annular serrations depress themselves against the inside wall of the bottle neck, making each serration a suction sealing point.
Note: For extra security when using large bore transfer needles, fasten septum to bottle or flask opening with a wrap-it tie. See ties and accessory tool listings on next page.
Note: For extra security when using large bore transfer needles, fasten septum to bottle or flask opening with a wrap-it tie. See ties and accessory tool listings on next page.
Features and Benefits
- Precision fit
- Non-contaminating
- Safety and performance
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service