Z503088
Threaded vial connector
20-400 thread for 22-400
Synonym(s):
747205-2022
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Recommended Products
fitting
20-400 thread for 22-400
Related Categories
General description
These threaded connectors provide a convenient method to connect standard screw thread sample vials to glassware with GPI threads and to standard taper joint adapters.
• Excellent for collecting samples from rotary evaporator traps
• Made of glass filled nylon with inert PTFE inner seals
• Temp. range: up to 200 °C
• Excellent for collecting samples from rotary evaporator traps
• Made of glass filled nylon with inert PTFE inner seals
• Temp. range: up to 200 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)