Key Documents
Z418846
Sigma-Aldrich
Aldrich® flat-bottom flask
capacity 2,000 mL, joint: ST/NS 24/40
Synonym(s):
bottom flask, flat bottomed flask, flat flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
flat bottom flask
glass
short
single-neck
Joint
joint: ST/NS24/40
description
short neck
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Sigma-Aldrich
capacity
2,000 mL
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)