Z341908
IKA® universal attachments for KS 130/260 basic and control shakers
AS 260.1 universal attachment
All Photos(1)
About This Item
UNSPSC Code:
41103811
NACRES:
NB.73
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
IKA
availability
available only in EU
Related Categories
General description
Note: This attachment is for all kinds of vessels; infinitely variable clamping rolls allow universal adaptation to the vessels.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)