Z336637
Heidolph mechanical stirrers
Model RZR 2020, AC/DC input 220 V AC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
torque <= 400 Ncm
feature
CE compliant
Analogue speed setting
manufacturer/tradename
Model RZR 2020
availability
not available in USA and Canada
output
27 watts
Looking for similar products? Visit Product Comparison Guide
General description
Highest torque of its class, two gear speeds. Mechanical speed setting and stepless transmission. Designed for continuous operation. Superior safety by overheat protection, motor cut-out feature in a permanent over-load situation. Encapsulated housing, slim-line space-saving design. Clear front panel layout for ease of operation. Maintenance and spark-free condenser motor. Includes chuck and 3 year warranty.
Speed range I 40 - 400rpm
Speed range II 200 - 2000rpm
Speed range I 40 - 400rpm
Speed range II 200 - 2000rpm
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service