Key Documents
Z300888
Quickfit® Reduction Adapters
top female joint: ST/NS 14/23, bottom male joint: ST/NS 24/29
Synonym(s):
DA13
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
female joint: ST/NS top14/23
male joint: ST/NS bottom24/29
manufacturer/tradename
Quickfit DA 13
availability
not available in USA and Canada
General description
These adapters connect items with a smaller cone to a piece of glassware with a larger size socket.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)