Z291994
WISP™ 96-position autosampler vials
volume 1 mL, clear glass vial, pkg of 250 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
clear glass vial
low-density polyethylene closure
feature
closure type polyethylene cap
packaging
pkg of 250 ea
O.D. × H
8 mm × 40 mm
volume
1 mL
fitting
plug for 8 mm
General description
8 × 40mm, 1mL vol. Constructed of clear borosilicate glass. Vials come with LDPE snap-plug caps.
Legal Information
WISP is a trademark of Waters
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service