Key Documents
Z286451
Nalgene® stopcock
3-way, bore size 2 mm
Synonym(s):
3-way connector, hose connector
About This Item
UNSPSC Code:
41100000
NACRES:
NB.79
Pricing and availability is not currently available.
material
polypropylene (with TFE plug)
description
for tubing I.D. 1/4 -5/16 in.
feature
autoclavable
packaging
pack of 1 ea
case of 4 ea
manufacturer/tradename
Nalgene® 6470, 0002
bore size
2 mm
Related Categories
General description
Serrated tubulation on each end.
Serrated tubulations arranged in a T-shape.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)