Skip to Content
Merck
All Photos(1)

Key Documents

Z276669

Sigma-Aldrich

UV quartz cuvette

Standard rectangular, chamber volume 3.5 mL, PTFE stopper

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
NB.44
Pricing and availability is not currently available.

material

PTFE stopper
quartz

manufacturer/tradename

Starna 21/Q/10/UM

parameter

170-2,700 nm spectral range

H × W × D

48 mm × 12.5 mm × 12.5 mm

chamber volume

3.5 mL

pathlength

10 mm

Looking for similar products? Visit Product Comparison Guide

General description

Spectrally matched Spectrosil® for UV quartz cells for quality control and research applications. Cuvette cells have a fused body with optically polished quartz windows. Fit all standard spectrophotometer cell holders.

Application



  • Enhanced detection and differentiation of THC isomers: A study developed an enhanced liquid chromatography-tandem mass spectrometry (LC-MS-MS) method for distinguishing 8- and 9-tetrahydrocannabinol isomers in blood and urine, utilizing synthetic analogs like (-)-11-nor-9-Carboxy-delta8-THC for accurate forensic toxicology and cannabinoid profiling (Reber et al., 2022).


Legal Information

Starna is a trademark of Starna Cells Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. ¿Las celdas de cuarzo son de boca ancha o con orificio circular?

    1 answer
    1. This product has a circular hole with a PTFE stopper. This is a product of StarnaCells. See the supplier drawing and specifications below.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service