Z261718
Luer-to-Closed end stopper
Micro-Mate® female Luer, 316 stainless steel
All Photos(1)
About This Item
UNSPSC Code:
41122500
NACRES:
NB.03
Recommended Products
material
316 stainless steel
manufacturer/tradename
Popper 6450
Related Categories
General description
Hardware specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes.
316 SS fittings are noncontaminating, easy to clean and resistant to corrosion. Chrome-plated brass connectors offer good performance at an economical price. Water tested at 45 psi.
316 SS fittings are noncontaminating, easy to clean and resistant to corrosion. Chrome-plated brass connectors offer good performance at an economical price. Water tested at 45 psi.
Legal Information
Micro-Mate is a registered trademark of Cadence Science
Choose from one of the most recent versions:
Certificates of Analysis (COA)