Key Documents
Z254231
Sigma-Aldrich
Filter Adapter Set
seven black neoprene rubber adapters, sizes 1 through 7
Synonym(s):
Adapter Set, Filter Adapter Kit
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
material
neoprene
rubber
manufacturer/tradename
Aldrich
Related Categories
General description
Set of seven black neoprene rubber adapters for Buchner, conical, fritted-glass, and porcelain filter funnels. One of each size 1 through 7. Withstands autoclaving.