Key Documents
Z243574
Sigma-Aldrich
Aldrich® dual bank manifold with glass stopcocks
positions, 5
Synonym(s):
Dual Bank Manifold, Glass Stopcock Manifold, Glass Stopcocks Manifold
About This Item
UNSPSC Code:
40161501
NACRES:
NB.79
feature
positions 5
L
500 mm
Related Categories
General description
replacement stopcock is Z681504
4 mm bore glass stopcocks.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)