Key Documents
Z218898
Ace giant extraction thimble
I.D. × L 60 mm × 180 mm
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
manufacturer/tradename
Ace Glass 681202
I.D. × L
60 mm × 180 mm
Related Categories
General description
For use with Ace giant Soxhlet extractors. Thimbles are readily permeable to the flow of ether and other organic solvents. Made of cellulose.
Choose from one of the most recent versions:
Certificates of Analysis (COA)