Skip to Content
Merck
All Photos(1)

Key Documents

Z218774

Ace giant Soxhlet extraction apparatus

flask capacity 12 L, extractor capacity 5 L, thimble I.D. × L 110 mm × 300 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

size

C

manufacturer/tradename

Ace Glass 681030

Overall H

82 in.

extractor capacity

5 L

flask capacity

12 L

thimble I.D. × L

110 mm × 300 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Complete with bulb type condenser and one flask. Cycling rate may be doubled over conventional extractors. All joints are interchangeable. Size C is supplied with adapter Z218871 for connecting the XL extractor joint and condenser. Order giant thimbles separately.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service