Z202762
Aldrich® adjustable pressure-release valve
pressure 5-50 psi
All Photos(1)
About This Item
UNSPSC Code:
40141600
NACRES:
NB.79
Recommended Products
parameter
5-50 psi pressure
Related Categories
General description
Automatic, adjustable pressure relief valve is for use on apparatus where internal pressure build-up is a concern. Can be mounted into a vacuum rack to allow rapid purging of an inert gas without damage to system. Also useful for freezing applications. Two-piece PTFE plug with Viton® O-ring and SS spring is adjustable for pressures of 5 to 50 psi. The tension on the spring determines the amount of pressure needed to force the lower part of the two-piece PTFE valve plug to open.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)