Key Documents
Z190276
Sigma-Aldrich
Arrow electric lab stirrer
speed 0-1750 rpm, direct, output0.066 hp
Synonym(s):
lab mixer, lab mixers, mixer stirrer
About This Item
UNSPSC Code:
41103801
NACRES:
NB.72
Pricing and availability is not currently available.
AC/DC input
115 V
115 V
packaging
pack of 1 ea
manufacturer/tradename
Arrow Engineering 1750
availability
not available in EU
parameter
0-1750 rpm speed
output
0.066 hp
General description
Totally enclosed, non-ventilated motor seals out corrosive fumes and dust. With on/off switch, circuit-breaker, and variable speed control. Includes a 9/16 × 10 in. support rod and 3/8 diam. × 12 in. L SS shaft and propeller.
Quality
Note: Arrow lab stirrers conform to CSA22.2 but are not certified
Choose from one of the most recent versions:
Certificates of Analysis (COA)