Z183415
Foil/poly bag
size 18 in. × 22 in.
Synonym(s):
Foil Bag, Poly Bag
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
24111500
NACRES:
NB.35
Recommended Products
packaging
pack of 10 × 30 bags
size
18 in. × 22 in.
thickness
5 mil
Looking for similar products? Visit Product Comparison Guide
General description
Heat sealable. Useful for storing air, light, or moisture-sensitive reagents, odoriferous products, and for temporarily containing leaking bottles.
Recommended heat seal conditions: 400°F (204°C), 40 psi, 1.5 sec.
Recommended heat seal conditions: 400°F (204°C), 40 psi, 1.5 sec.
Packaging
1PAK ≡ 10 bags
30PAK ≡ 300 bags
30PAK ≡ 300 bags
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service