Key Documents
Z180939
Ace auto-zero McLeod vacuum gauges
pressure 0-5 mmHg
About This Item
UNSPSC Code:
41112406
NACRES:
NB.79
manufacturer/tradename
Ace Glass 872549
pressure measuring range
0-5 mmHg
Related Categories
General description
A tilting gauge with a constant zero overflow which eliminates the capillary-depression error from an expanding meniscus. Once the zero line of the scale is aligned with the tip of the closed capillary, it is only necessary to bring the gauge to an upright position and read the scale behind the closed leg.
Gauges are fabricated from calibrated ACE Trubore tubing, to assure uniform and reproducible reading between the gauges. Mercury trap is incorporated in female joint, bulbs are enlarged. Joint is standard taper 20/65.
The pressure range is: 0-1.0 mmHg and the lowest reading is: 0.001 mmHg.
Mercury not included (approximately 20 mL of triple distilled mercury is needed to ffill this vacuum gauge to working volume).
Choose from one of the most recent versions:
Certificates of Analysis (COA)