Key Documents
Z177857
Sigma-Aldrich
Scienceware® Ellipso-Spoon® sampler
L 18 cm
Synonym(s):
curved laboratory spatula, laboratory spoon
About This Item
UNSPSC Code:
41122403
NACRES:
NB.35
Pricing and availability is not currently available.
manufacturer/tradename
Bel-Art H368060018
L
18 cm
General description
Special curved shape permits contact with all corners of vessels, bottles, or containers. No corner out of reach and no loss of materials. Handles crystals, powders, granulated and solid substances, even thick liquids or delicate substances. SS, will not bend. Has both spoon end and spatula end.
Legal Information
Ellipso-Spoon is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)