The jacketed beaker has approximately 75 mm internal diameter, 120 mm internal depth (depth of inner well) and 140 mm height.
Recommended Products
material
borosilicate glass
feature
graduations: no
volume
400 mL
Related Categories
General description
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
-
HI! COULD YOU PLEASE DETAIL THE INSIDE DIAMETER OF THE BEAKER?
1 answer-
Helpful?
-