Key Documents
Z125679
Sigma-Aldrich
Ace three-neck round-bottom flasks
vertical, capacity 1,000 mL, center joint: ST/NS 29/42, side Joints: ST/NS 24/40 (2)
Synonym(s):
bottom flask, flask round, round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass
round bottom flask
three-neck (Vertical)
Joint
center joint: ST/NS29/42
side Joints: ST/NS24/40 (2)
sterility
non-sterile
feature
closure type Standard Taper joint
packaging
pack of 1
manufacturer/tradename
Ace Glass 6944-26
capacity
1,000 mL
Choose from one of the most recent versions:
Certificates of Analysis (COA)