Skip to Content
Merck
All Photos(1)

Key Documents

Z125318

Ace PTFE ferrules

for tubing o.d., 3.2 mm (1/8 in.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
27121703
NACRES:
NB.35

Pricing and availability is not currently available.

feature

for tubing o.d. 3.2 mm (1/8 in.)

manufacturer/tradename

Ace Glass 1171003

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
COHISR-ROSAB1306082HPPCRPKRO
manufacturer/tradename

Roche

manufacturer/tradename

Roche

manufacturer/tradename

-

manufacturer/tradename

Roche

parameter

1,420 cm/hr max. flow rate

parameter

-

parameter

-

parameter

-

capacity

≥40 mg/mL, resin binding capacity

capacity

-

capacity

-

capacity

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

-

General description

For use in place of O-rings on threaded glassware.

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service