material
round hole
technique(s)
IR spectroscopy: suitable
diam.
1.5 mm
General description
For making smaller KBr pellets. Use with the Aldrich macro-micro KBr pellet die (Z506699). Inserts also fit the Magnetic KBr pellet holder (Z112489) or Econo-Card (Z123544).
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)