Key Documents
Z116645
Sigma-Aldrich
Aldrich® gas-condensing bottle
with Luer lock
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
material
cap (Luer lock)
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
150 mL
Related Categories
General description
With stopcock, graduated, holds 150 mL of liquefied gas.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)