Y906468
Valve
stainless steel diaphragm, for CGA 660 right angle path, external, right hand thread
All Photos(1)
About This Item
UNSPSC Code:
12142200
NACRES:
NA.12
Recommended Products
material
stainless steel diaphragm
Quality Level
parameter
2640 psi max. pressure
L
4.62 in.
weight
560 g
fitting
for CGA 660 right angle path, external, right hand thread
Related Categories
Other Notes
This product is only available with the purchase of a bulk gas product and not for individual sale. For more information, please contact Stable Isotopes Customer Service
Choose from one of the most recent versions:
Certificates of Analysis (COA)