WHAWB100035
Qiagen® swabs for bucal and saliva sample collection
Omni Swab, sterile, pkg of 100 ea
Synonym(s):
OmniSwab, Sterile (100), Whatman fta, Z747394, fta Whatman, omni swab, omniswab
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
sterility
sterile
packaging
pkg of 100 ea
manufacturer/tradename
Qiagen WB100035
application(s)
environmental
food and beverages
suitability
bacteria
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Sterile collection swabs with ejectable brush-like head used for buccal and saliva samples and convenient DNA extraction.
Features and Benefits
- Omni Swabs are manufactured with an absorbent material that increases buccal cell concentrations to give high quality results.
- Compatible with standard STR amplification chemistry.
- OmniSwab collection swabs are individually packaged in a clean-room environment and sterilized by gamma irradiation.
Other Notes
Field of Use : For internal research use only
Legal Information
Qiagen is a registered trademark of Qiagen Group
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service