WHA68941304
Whatman® GD/X syringe filters
glass microfiber membrane, pore size 0.45 μm, diam. 13 mm, non-sterile, pk of 150 ea
Synonym(s):
Whatman filter, Z277339, syringe filter, syringe filters
About This Item
Recommended Products
material
polypropylene housing
polypropylene membrane (pigment-free)
sterility
non-sterile
feature
glass fiber prefilter
packaging
pk of 150 ea
manufacturer/tradename
Whatman 6894-1304
Whatman Article No. 28420545 (US reference)
diam.
13 mm
volume
≤10 mL
pore size
0.45 μm
fitting
female Luer inlet (lock)
male Luer outlet
Related Categories
General description
Features and Benefits
- Non sterile Whatman GD/X Syringe Filters combine a polypropylene housing, high-efficiency Whatman GMF 150 and GF/F pre-filter media, and one of the following membrane materials:
- Polyethersulfone (PES) for serum, plasma and tissue culture
- Polyvinylidene difluoride (PVDF) for HPLC
- Cellulose acetate for biological solutions
- Borosilicate glass microfiber
Suitability
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)